Check Order Status
Cryo Gloves - Mid-Arm Length
Actual product may vary
Cryo Gloves - Mid-Arm Length
Safely handle samples from ultra-low and cryogenic storage with these cryo gloves which offer a high level of thermal protection.
Have Questions?
Change view
Catalog Number
Size
189561
also known as GT189561
Medium
189440
Small
189562
also known as GT189562
Large
3 Options
Catalog number
189561
also known as GT189561
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Size:
Medium
Safely handle samples from ultra-low and cryogenic storage with these cryo gloves which offer a high level of thermal protection.
Comfortable, lightweight, and breathable protection
Side release buckles for quick on/off
Available in a variety of sizes
Specifications
Description
Mid-Arm Length
Color
Blue
Hand
Specific
Size
Medium
For Use With (Application)
Handling Samples from Ultra Low Temperature and Cryogenic Storage
Length (English)
14 in.
Length (Metric)
35.6 cm
Unit Size
PAA