Check Order Status
Cryo Gloves - Mid-Arm Length
Actual product may vary
Cryo Gloves - Mid-Arm Length
Safely handle samples from ultra-low and cryogenic storage with these cryo gloves which offer a high level of thermal protection.
Have Questions?
Change view
Catalog Number
Size
189440
Small
189561
Medium
189562
Large
3 Options
Catalog number
189440
Price (MYR)
2,264.00
PAA
Add to cart
Size:
Small
Price (MYR)
2,264.00
PAA
Add to cart
Safely handle samples from ultra-low and cryogenic storage with these cryo gloves which offer a high level of thermal protection.
Comfortable, lightweight, and breathable protection
Side release buckles for quick on/off
Available in a variety of sizes
Specifications
Description
Mid-Arm Length
Color
Blue
Hand
Specific
Size
Small
For Use With (Application)
Handling Samples from Ultra Low Temperature and Cryogenic Storage
Length (English)
14 in.
Length (Metric)
35.6 cm
Unit Size
PAA