Van Goat Cut A6 with Gauntlet
Van Goat Cut A6 with Gauntlet
Actual product may vary
Van Goat Cut A6 with Gauntlet
Van Goat Cut A6 with Gauntlet is made of Cutshield™ Para-aramid/steel/polyester liner for superior ANSI A6 protection.
Have Questions?
Catalog NumberSize
171-00011
also known as PAI171-00011
2X-Large
Catalog number 171-00011
also known as PAI171-00011
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Size:
2X-Large
  • Full sock Cutshield™ Para-aramid/steel/polyester liner
  • Dryhide oil and Water Resistant full-grain goatskin leather
  • Gunn cut
  • Wing thumb
  • Pulse protector
  • Gauntlet style cuff
Specifications
Certifications/ComplianceANSI Cut, ANSI Puncture
Certifications/Compliance (Europe)EN388
Coated AreaUncoated
ConstructionCut and Sewn
Cuff MaterialLeather Gore
Coating MaterialUncoated
ColorCream
Cut Resistance RatingANSI A6
HandAmbidextrous
Latex FreeYes
MaterialGoatskin Leather
Product TypeCut-resistant Gloves
Size2X-Large
EN388 StandardEN388:2X33F
For Use With (Application)Oil and Gas, LNG, Construction
GenderMen's
Glove Cuff ColorGray
Glove Cuff StyleGauntlet
Liner MaterialUnlined
LiningUnlined
Model549
Puncture RatingANSI/ISEA 105-16 Level 4
SeriesVan Goat
Length (English)12 in.
Length (English) Cuff5 in.
Length (Metric)30 cm
Length (Metric) Cuff13 cm
Unit SizeEA