Perform microbiological QC testing and analysis of water, food/beverage, raw material and finished product with these economical sterile disposable funnels.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Pore Size
Color
147-2045
250 mL
0.45 μm
Gray with Black Grid
140-4045
150 mL
0.45 μm
White with Gray Grid
2 Options
Catalog number 147-2045
Price (MYR)
2,111.00
CS
Add to cart
Capacity (Metric):
250 mL
Pore Size:
0.45 μm
Color:
Gray with Black Grid
Price (MYR)
2,111.00
CS
Add to cart
Use these economical sterile disposable filter funnels to perform microbiological QC testing and analysis of water, food/beverage, raw material and finished product. The squeeze and twist funnel is easy to remove for retrieval of the CN membrane.
Features:
High impact polystyrene collars are rugged for reliable performance
Preassembled, sterile, individually bagged to ensure sample integrity
Adapters accommodate No. 8 stoppers for use with 0.5 in. hole in vacuum manifolds or filter flasks
47mm diameter cellulose nitrate membrane is certified for water quality work
250mL sizes are useful when testing beverages that may foam
Cat. No. 147-0045 features gray membrane that turns black when wet, making it easier to detect and count growing colonies
Note: Designed for single use only
Includes: Six funnel adapters with each case
Specifications
Capacity (Metric)250 mL
Certifications/ComplianceMeet requirements of APHA Standard Methods and EPA requirements for water quality work.
ColorGray with Black Grid
Diameter (Metric) Filter47 mm
For Use With (Equipment)Filter Manifold, Filtering Flask