Bosshead
Product Image
Bosshead
Available as either fixed or rotatable.
Have Questions?
Catalog NumberSize (Metric) GripDescription
1388
also known as 1388-FMT
25 mmHeavy duty, with fixed right angle Vee grooves
Catalog number 1388
also known as 1388-FMT
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Size (Metric) Grip:
25 mm
Description:
Heavy duty, with fixed right angle Vee grooves
Bossheads available as either fixed or rotable, with varying opening diameters. Economy type available.
  • Bosshead
  • To hold objects of various diameters
  • Both fixed and rotatable options available
Specifications
QuantityEach
TypeBosshead Clamp
For Use With (Equipment)Hold up to 25 mm Diameter Object
For Use With (Application)To Hold up to 25 mm Diameter Objects
DescriptionHeavy duty, with fixed right angle Vee grooves
Size (Metric) Grip25 mm
Unit SizeEA