Plain Microscope Slides
Plain Microscope Slides
Actual product may vary
Plain Microscope Slides
Globe Scientific Inc. Plain Microscope Slides produced from premium glass sheets, available in 3 different styles
Have Questions?
Catalog NumberCorner StyleEdges
1331ClippedBeveled
Catalog number 1331
HSN 84213990
Price (MYR)
0.00
EA
Add to cart
Corner Style:
Clipped
Edges:
Beveled
Price (MYR)
0.00
EA
Add to cart
  • Offer exceptional quality while providing considerable savings
  • Slides are pre-cleaned and ready for use
Specifications
No. per Case1440
No. per Pack72
SterilityNon-sterile
EdgesBeveled
FrostedNot Frosted
Length (Metric)75 mm
MaterialSoda Lime Glass
PrecleanedYes
Thickness (Metric)1.1 mm
Width (Metric)25 mm
ColorClear
Corner StyleClipped
Dimensions (L x W)75 x 25 mm
Unit SizeEA