2-Aminobenzonitrile, 98%
2-Aminobenzonitrile, 98%
2-Aminobenzonitrile, 98%
2-Aminobenzonitrile, 98%
Acros Organics
2-Aminobenzonitrile, 98%
CAS: 1885-29-6 | C7H6N2 | 118.14 g/mol
Have Questions?
Catalog NumberQuantityPackaging
104921000
also known as 10492
100 gGlass bottle
Catalog number 104921000
also known as 10492
HSN 29269000
Price (MYR)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1885-29-6
IUPAC Name2-aminobenzonitrile
Molecular FormulaC7H6N2
InChI KeyHLCPWBZNUKCSBN-UHFFFAOYSA-N
SMILESNC1=CC=CC=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to yellow to beige to brown
Infrared spectrumConforms
Appearance (Form)Powder or crystals or flakes and/or chunks
Melting point46°C to 51°C
GC>=97.5 %