Single-Cell Sample Holders for Evolution 300 and 600 UV-Visible Spectrophotometers
Single-Cell Sample Holders for Evolution 300 and 600 UV-Visible Spectrophotometers
Actual product may vary
Single-Cell Sample Holders for Evolution 300 and 600 UV-Visible Spectrophotometers
For use with Thermo Scientific™ Evolution™ 300 and 600 spectrophotometers
Have Questions?
Catalog NumberHolds
10011501
also known as FA100115C01
One rectangular cell
Catalog number 10011501
also known as FA100115C01
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Holds:
One rectangular cell
Adjustable Microcell Holder
Holds one ultra-microcell, microcell or standard rectangular cell with 10mm maximum pathlength
Cylindrical Cell Holder
Holds one cylindrical cuvet with 100mm maximum pathlength
Long Pathlength Rectangular Cell Holder
Holds one rectangular cuvet with 5 to 100mm pathlength
Test Tube and Vial Holder
Holds one test tube or vial cuvet with diameter from 10 to 16mm and height from 75 to 100mm
Specifications
TypeMicrocell Holder
For Use With (Equipment)Thermo Scientific™ Evolution™ 300 and 600 UV-visible spectrophotometer
HoldsOne rectangular cell
IncludesOne cuvet holder on base plate
Pathlength10 mm
Unit SizeEA