Fiberlite™ H3-LV Large Volume Swinging Bucket Rotor
Fiberlite™ H3-LV Large Volume Swinging Bucket Rotor
Actual product may vary
Fiberlite™ H3-LV Large Volume Swinging Bucket Rotor
Conduct high-volume microplate processing with this large volume swinging bucket rotor.
Have Questions?
Catalog NumberFor Use With (Equipment)
096-029027Beckman GS-6, Allegra 6, GP Series Centrifuges
Catalog number 096-029027
Price (MYR)
-
For Use With (Equipment):
Beckman GS-6, Allegra 6, GP Series Centrifuges
Conduct high-volume microplate processing with the Thermo Scientific™ Fiberlite™ H3-LV Large Volume Swinging Bucket Rotor.
Fiberlite rotors:
  • Lightweight carbon fiber rotors reduce run-times by achieving higher g-forces
  • Corrosion-and fatigue-resistant design ensures structural integrity.
  • Rotors are backed by a 15-year warranty and are compatible with Thermo Scientific™ Nalgene™ bottles and tubes for complete centrifuge solutions.
Recommended for:
Pelleting cells and cellular debris, protein precipitation, plasmid purification and collecting physiological fluids for diagnostic tests.
Note:
Performance may vary by centrifuge model.
Specifications
DescriptionFiberlite H3-LV Large Volume Swinging Bucket Rotor for Beckman GS-6, Allegra 6, GP Series
TypeSwinging Bucket Rotor
Capacity28 Standard or 8 Deepwell Plates
Max. RCF1,840 x g
Max. Speed3,600 rpm
For Use With (Equipment)Beckman GS-6, Allegra 6, GP Series Centrifuges
Angle90°
Unit SizeEA