Check Order Status
1/4 in. Graphite Fittings
Actual product may vary
1/4 in. Graphite Fittings
Provide leak-free system connections. Trajan Scientific and Medical 1/4 in. Graphite Fittings leads to improved chromatography for non-MS systems.
Have Questions?
Catalog Number
Diameter (Metric) Inside
072617
5/16 in.
Catalog number
072617
Price (MYR)
-
Diameter (Metric) Inside:
5/16 in.
Easy to use
Forms reliable seal
Reusable
Soft material, forms soft grip to the capillary column
Low emissions
Specifications
Size (Metric)
9.5 mm x 4.6 mm
Size (Metric) Fitting
1/4 in.
Material
Graphite
Type
Graphite Reducing Ferrule 1/4 in. Fitting 5/16 in. x 9.5mm x 4.6mm
Quantity
10/Pk.
Diameter (Metric) Inside
5/16 in.