Wash Kit 10 mL Snap Vial Silicone/PTFE Septa 25 pcs
Wash Kit 10 mL Snap Vial Silicone/PTFE Septa 25 pcs
Actual product may vary
Wash Kit 10 mL Snap Vial Silicone/PTFE Septa 25 pcs
Wash Kit 10 mL Snap Vial Silicone/PTFE Septa 25 pcs
Have Questions?
Catalog NumberColor
WSKT-25White
Catalog number WSKT-25
Price (MYR)
-
Color:
White
Wash Kit 10 mL Snap Vial Silicone/PTFE Septa 25 pcs
Wash Kit 10 mL Snap Vial Silicone/PTFE Septa 25 pcs
Specifications
Diameter (English) Outer0.79 in.
ColorWhite
Diameter (Metric) Outer20 mm
SterilitySterile
Volume (Metric)10 mL
For Use With (Application)Chromatography
No. per Pack25 Pk.
Volume (English)0.34 oz.
Closure MaterialPTFE, Silicone
AutoclavableAutoclavable
Closure TypeSnap Cap
TypeSample Vial Kit
Closure Size20 mm
Unit SizePack