Wheaton™ Amber Wide-Mouth Packers with PTFE Faced PP-lined Closures
Wheaton™ Amber Wide-Mouth Packers with PTFE Faced PP-lined Closures
Actual product may vary
Wheaton™ Amber Wide-Mouth Packers with PTFE Faced PP-lined Closures
Amber glass for storing light-sensitive powders
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
WHEW21694530 mL28-400 mm
FIS02-912-15530 mL28-400 mm
FIS02-912-145250 mL45-400 mm
FIS02-912-1471000 mL53-400 mm
Catalog number WHEW216945
Price (MYR)
-
Capacity (Metric):
30 mL
Closure Size:
28-400 mm
Amber glass for storing light-sensitive powders
  • USP Type III soda-lime glass
  • Allows for easy removal of contents
Specifications
Capacity1 oz., 30mL
Capacity (Metric)30 mL
Capacity (English)1 oz.
ColorAmber
Diameter (English)1.45 in.
MaterialSoda Lime Glass
Closure ColorWhite
Certifications/ComplianceUSP Type III
Closure IncludedYes
MouthWide
ShapeRound
Height (Metric)67 mm
Product TypePacker Bottle
Height (English)2.63 in.
Closure LinerPTFE/Polypropylene
Closure MaterialPolypropylene
Closure Size28-400 mm
Diameter (Metric)37 mm
Closure TypeScrew Cap
Through the Closure DispensingYes
Unit SizeCS