Check Order Status
Oil Mist Separator for CRVpro Direct Drive Rotary Vane Vacuum Pump
Actual product may vary
Oil Mist Separator for CRVpro Direct Drive Rotary Vane Vacuum Pump
For use with CRVpro Direct Drive Rotary Vane Vacuum Pump
Have Questions?
Catalog Number
For Use With
WEL320015
Rotary vane pump
Catalog number
WEL320015
Price (MYR)
0.00
EA
Add to cart
For Use With:
Rotary vane pump
Price (MYR)
0.00
EA
Add to cart
Outlet oil separator removes oil particles from exhaust stream, minimizing oil loss to atmosphere
Specifications
Description
Outlet Oil Mist Separator
For Use With (Application)
Removes oil particles from exhaust stream
For Use With
Rotary vane pump