Check Order Status
NS 1643
Element Search
Structure Search
NS 1643
KV11.1 (hERG) channel activator; antiarrhythmic
Have Questions?
Catalog Number
Quantity
WEL3062-10
10 mg
Catalog number
WEL3062-10
Price (MYR)
22,724.00
EA
Add to cart
Quantity:
10 mg
Request bulk or custom format
Price (MYR)
22,724.00
EA
Add to cart
Specifications
Chemical Name or Material
NS 1643
CAS
448895-37-2
Recommended Storage
Store at +4°C
Quantity
10 mg
Molecular Formula
C15H10F6N2O3
View more
Human ether-a-go-go related gene (hERG) KV11.1 channel activator (EC50 = 10.5 μM). Exhibits different molecular mechanisms of action at KV11.1 (hERG1) and KV11.2 (hERG2) channels. Displays antiarrhythmic activity
in vitro
.