WOB-L Piston Dry Vacuum Pump 2534
WOB-L Piston Dry Vacuum Pump 2534
Actual product may vary
WOB-L Piston Dry Vacuum Pump 2534
Perfect for many common laboratory vacuum applications. Welch™ WOB-L Piston Dry Vacuum Pump 2534 is reliable, easy to adjust, and easy to monitor.
Have Questions?
Catalog number WEL2534C-02
Price (MYR)
-
  • Economical
  • Oil free operation for reduced maintenance
  • High water vapor tolerance
  • Inlet liquid trap
  • Adjustable Vacuum / Pressure
  • Applications: Filtration (1 to 4 funnels), aspiration, desiccation, SPE, replace water aspirator, vacuum drying
Specifications
Max. Vacuum27.2 in.Hg
IncludesVacuum (and pressure) regulator, vacuum (and pressure) gauge, silencer and water trap
Electrical Requirements230 V 50 Hz 1 Ph2.
Dimensions (L x W x H)20.6 x 22 x 25.4 cm
Plug TypeEuropean (Schuko)
Wattage93 w
Pump TypeDry Vacuum Pump
Air Displacement34 L/min.
ApplicationsFiltration (1 to 4 funnels), aspiration, desiccation, SPE, replace water aspirator, vacuum drying
Maximum Pressure100 psig