Ethanol Absolute Ph.Eur., Solveco
Ethanol Absolute Ph.Eur., Solveco
Ethanol Absolute Ph.Eur., Solveco
Absolute, pharmaceutical-grade ethanol with a concentration of 99.9 vol - %. This synthetic ethanol is produced through a complex three-step process, resulting in a double-distilled ethanol.
Have Questions?
Catalog NumberQuantity
WEL101725 L
Catalog number WEL1017
Price (MYR)
-
Quantity:
25 L
Request bulk or custom format
Specifications
Chemical Name or MaterialEthyl Alcohol
CAS64-17-5
Boiling Point78°C
Quantity25 L
PackagingHDPE Container
View more
A synthetic ethanol that is produced through a complex three-step process, resulting in double-distilled ethanol. This makes it an ideal product for applications requiring the highest quality ethanol. Synthetic ethanol is manufactured using selective synthetic chemistry, which eliminates risks associated with contaminants such as genetically modified organisms, pesticide residues, natural allergens, phthalates, and dioxins. With a purity of 99.9%, this ethanol meets the quality requirements of the European Pharmacopoeia and is available with a batch-specific Certificate of Analysis upon request.