OSOM™ Ultra Flu A and B Test Control Swab Kit
OSOM™ Ultra Flu A and B Test Control Swab Kit
Actual product may vary
OSOM™ Ultra Flu A and B Test Control Swab Kit
OSOM™ Ultra Flu A and B Test Control Swab Kit is a rapid qualitative test that detects influenza type A and type B antigens directly from nasal swab, nasopharyngeal swab and nasal aspirate/wash specimens.
Have Questions?
Catalog NumberQuantity
WEL100525 Pk.
Catalog number WEL1005
Price (MYR)
-
Quantity:
25 Pk.
Improved Outcomes: Rapid and accurate diagnosis of Influenza enables a choice of antiviral therapy, helps avoid the overuse of antibiotics, and prevents healthcare costs related to unnecessary testing and treatment.
Patient Experience: Streamlines lab's workflow while reducing patient wait time and unnecessary use of antibiotics.
  • For the detection of Influenza type A and type B nucleoprotein antigens
  • Able to meet FDA reclassification standards
  • Multiple specimen types: nasal swab, nasopharyngeal swab and nasal aspirate/wash specimens
  • Cleared for use with multiple viral transport media (VTM) types
  • Accuracy: Influenza A Sensitivity: >90%; Influenza B: >85% vs culture (across multiple sample types)
  • Fast: Obtain positive results in 10 to 15 minutes
  • Flexible: Runs from nasal swab, NP swab as well as nasal aspirate/wash
  • Workflow: approved for use with multiple viral transport media types
  • QC Inside: 2 extra no charge cassettes to the kit
OSOM Ultra Flu A and B Control Kit (Cat. No. 22321114): 5 x positive Flu A/B swabs and 5 x Negative Flu A/B swabs
OSOM Ultra Flu A and B Nasopharyngeal Wash/Aspirate Kit (Cat. No. 22321106): 50 pipets and 5 mL extraction reagent
OSOM Ultra Flu A and B Swab Transport Kit (Cat. No. 22321107): 25 swabs in transport tubes
Specifications
Certifications/ComplianceCleared for use with multiple viral transport media (VTM) types
CPT CodeFlu A: 87804; Flu B: 87804-59 (modifier used to indicate separate test)
Detectable AnalytesInfluenza A and B
Sample TypeNasal Aspirate/Wash, Nasal Swab, Nasopharyngeal Swab
SensitivityInfluenza A: >90%, Influenza B: >80% vs Culture
TypeOSOM Ultra Flu A and B Test Control Swab Kit
Includes5 x positive Flu A/B swabs and 5 x Negative Flu A/B swabs
Quantity25 Pk.
Test Time10 to 15 min.
Unit SizeEA
Contents & Storage
Room Temperature