Check Order Status
U.S. Stainless Steel Test Sieve Covers
Actual product may vary
U.S. Stainless Steel Test Sieve Covers
Sieve cover for Fisherbrand U.S. standard test sieve, Stainless Steel fits 8 in. dia. (20.3cm) sieves
Have Questions?
Catalog Number
Diameter (English) Outer
TYL8497
8 in.
Catalog number
TYL8497
Price (MYR)
455.00
EA
Add to cart
Diameter (English) Outer:
8 in.
Price (MYR)
455.00
EA
Add to cart
Fit all 8 and 12 in. diameter (20.3 and 30.5cm) sieves.
Specifications
Description
Top Cover With Recessed Ring
Type
Sieve Cover
Diameter (English) Outer
8 in.
Material
Stainless Steel
For Use With (Equipment)
8 in. dia. Test sieve
Includes
Recessed Ring
Diameter (Metric) Outer
203 mm