Check Order Status
B-5 Fixative (Stock), Sodium Acetate-Mercuric Chloride, Ricca Chemical
Element Search
Structure Search
B-5 Fixative (Stock), Sodium Acetate-Mercuric Chloride, Ricca Chemical
Have Questions?
Catalog Number
Quantity
TYL8491
4 L
Catalog number
TYL8491
Price (MYR)
566.00
EA
Add to cart
Quantity:
4 L
Request bulk or custom format
Price (MYR)
566.00
EA
Add to cart
Specifications
Physical Form
Solution
CAS
7732-18-5
CAS
7487-94-7
Packaging
Natural Poly Bottle
CAS
6131-90-4
View more