Camatril™ 730 Gloves
Camatril™ 730 Gloves
Actual product may vary
Camatril™ 730 Gloves
Camatril™ 730 Gloves
Camatril™ 730 Gloves
Honeywell KCL™ Camatril™ 730 Gloves are a top-quality protective glove made of high-quality nitrile, making them useful for a wide range of applications.
Have Questions?
Catalog NumberSize
TYL73099
Catalog number TYL7309
Price (MYR)
1,089.00
EA
Add to cart
Size:
9
Price (MYR)
1,089.00
EA
Add to cart
  • Top-quality nitrile
  • Large spectrum of chemical resistance
  • Versatile glove suitable for numerous fields
  • Good flexibility
  • Anatomical design and excellent fit
  • Good sensitivity
  • Flock lining for added comfort
  • Approved for food handling and processing according to EU 1935/2004
Specifications
Certifications/Compliance (Europe)EC Category PPE 3, suitable for food handling according to EU 1935/2004
Coating MaterialNitrile
ColorGreen
Cut Resistance Rating0
MaterialNitrile
Size9
Thickness (Metric)0.4 mm
LiningFlock Lined
Puncture Rating1
Resistant ToResistance to permeation by chemicals: AJL
Tear Rating0
Length (Metric)300-320 mm
Unit SizeEA