Eco Sleeve Guard Dispenser, Double, Clear Recycled Plastic with Hinged Lid
Eco Sleeve Guard Dispenser, Double, Clear Recycled Plastic with Hinged Lid
Actual product may vary
Eco Sleeve Guard Dispenser, Double, Clear Recycled Plastic with Hinged Lid
High quality dispenser.
Have Questions?
Catalog number TYL7307
Price (MYR)
1,089.00
EA
Add to cart
Price (MYR)
1,089.00
EA
Add to cart
ZING Sleeve Guard Dispenser conveniently stores sleeve guards for quick and easy access. Sleeve Guard Dispenser is made from heavy-duty 100% recycled PETG providing superior durability and crack resistance over other materials like acrylic. Dispenser is ideal for high use applications and can be mounted on a wall or placed on a counter-top. Durable PETG hinges support a lid that keeps contents clean and dust free. Recycled materials support sustainability initiatives and LEED Credits. Sleeve Guard Dispenser uses clear plastic with 2 compartments and includes pre-drilled holes for easy mounting. Measures 12 in.H x 11.5 in.W x 10.5 in.D. Sleeve Guards Not Included. Made in the USA.
Specifications
Product TypeDispenser
ColorClear
Dimensions (L x W)11.5 x 10.5 in.
Width (English)10.5 in.
Height (English)11.5 in.
MaterialPlastic
MountingCorner Holes
For Use With (Application)Dispenser
Quantity1 Ea.
Unit SizeEA