Eco Bouffant Cap Dispenser, Single Box, Clear Recycled Plastic
Eco Bouffant Cap Dispenser, Single Box, Clear Recycled Plastic
Actual product may vary
Eco Bouffant Cap Dispenser, Single Box, Clear Recycled Plastic
High quality dispenser.
Have Questions?
Catalog number TYL7305
Price (MYR)
1,089.00
EA
Add to cart
Price (MYR)
1,089.00
EA
Add to cart
ZING Bouffant Cap Dispenser conveniently stores and organizes bouffant boxes on a wall in either the vertical or horizontal position. Bouffant Dispenser is made from heavy-duty 100% recycled PETG providing superior durability and crack resistance over other materials like acrylic. Dispenser is designed to hold one bouffant box up to 9-3/8 in. wide by 3-3/8 in. deep (bouffant box not included). Dispenser is ideal for high use applications and supports sustainability initiatives and LEED Credits. Includes pre-drilled holes for easy mounting. Overall measures 9 in.H x 10 in.W x 4 in.D. Made in the USA.
Specifications
Product TypeDispenser
ColorClear
Dimensions (L x W)10 x 4 in.
Width (English)4 in.
Height (English)10 in.
MaterialPlastic
MountingCorner Holes
For Use With (Application)Dispenser
Quantity1 Ea.
Unit SizeEA