RecycLockout Lockout Tagout, Forklift Propane Tank
RecycLockout Lockout Tagout, Forklift Propane Tank
Actual product may vary
RecycLockout Lockout Tagout, Forklift Propane Tank
Hollow cylindrical shell of 100% recycled polycarbonate that surrounds LP gas cylinder handwheels with a padlock (sold separately).
Have Questions?
Catalog NumberColor
TYL7293Yellow
Catalog number TYL7293
Price (MYR)
1,089.00
EA
Add to cart
Color:
Yellow
Price (MYR)
1,089.00
EA
Add to cart
ZING RecycLockout Forklift Propane Tank Lockout, locks out LP gas cylinder handwheels up to 2-3/16 in. diameter, accommodates 3 padlocks with shackles up to 9/16 in. diameter, 100% recycled yellow HDPE with highly visible label  in.DO NOT OPERATE in., UL Validated, supports sustainability initiatives and LEED credits, measures 2-1/2 in. Dia. x 3-1/2 in.W x 3-3/8 in.H, Made in the USA. 
Specifications
Product TypeLockout
ColorYellow
MaterialPlastic
DescriptionBracket mounting
Dimensions (L x W)3.5 x 3.5 in.
Height (English)3.5 in.
Quantity1 Ea.
Width (English)3.5 in.
Unit SizeEA