Corning microcarriers are designed to simplify your scale-up processes.
Have Questions?
Catalog Number
Quantity
TYL7289
5 g
Catalog number TYL7289
Price (MYR)
1,089.00
EA
Add to cart
Quantity:
5 g
Price (MYR)
1,089.00
EA
Add to cart
Sterile
Ready-to-use
Available in closed systems packaging to be used directly with bioreactors
Additionally, the microcarriers are surface-treated to enhance cell attachment, maximizing cell yield and viability
Consistency – USP Class VI polystyrene material provides a consistent platform
Performance – Corning microcarriers are offered untreated or with the following surface treatments
Corning Synthemax™ II substrate creates a synthetic surface on the microcarriers for stem cell expansion
For use with MSC and iPS/ES applications. (Animal Origin Free)
Sterility Assurance Level (SAL) of 10−6
Nonpyrogenic
Available as closed system solutions
Corning microcarriers are available sterile and ready to use in 1 to 10 g vials. This format provides flexibility to select the amount of beads to use.
During the mature phase of process development and into manufacturing, closed system packaging would be necessary to maintain sterility and ease bioreactor loading.
Specifications
For Use With (Application)MSC and iPS/ES Applications