HDPE T-Nipples for Pipes
HDPE T-Nipples for Pipes
Actual product may vary
HDPE T-Nipples for Pipes
These T-hose connectors are made of fracture-proof and food-safe HDPE with sleeve connections for hoses.
Have Questions?
Catalog NumberTubing Size
TYL537110mm
Catalog number TYL5371
Price (MYR)
956.00
EA
Add to cart
Tubing Size:
10mm
Price (MYR)
956.00
EA
Add to cart
  • Natural-colored HDPE
  • Very resistant to acids, alkalis and other chemicals
  • Available in different sizes
  • Temperature resistant from −20° to +70°C
Specifications
MaterialHDPE
For Use With (Application)Connecting hoses
Min. Operating Temperature (Metric)-20°C
Resistance PropertiesChemicals, Fractures
Tubing Size10mm
ColorNatural
Max. Operating Temperature (Metric)70°C
Product TypeT-nipple Hose Connection
Unit SizeEA