Platelet Aggregation Kit
Platelet Aggregation Kit
Actual product may vary
Platelet Aggregation Kit
Platelet Aggregation Kit is used to identify normal and abnormal platelet function, to quantitate platelet response and to determine the patient's response to anti-platelet drug therapy
Have Questions?
Catalog NumberSample Type
TYL5369Plasma
Catalog number TYL5369
Price (MYR)
956.00
EA
Add to cart
Sample Type:
Plasma
Price (MYR)
956.00
EA
Add to cart
Platelet Aggregation Kit is used to identify normal and abnormal platelet function, to quantitate platelet response and to determine the patient's response to anti-platelet drug therapy
  • The Platelet Aggregation Kit is used to identify normal and abnormal platelet function, to quantitate platelet response and to determine the patient's response to anti-platelet drug therapy
  • The reagents are optimised for use with the Helena AggRAM analyser, but may be used on any turbidometric aggregometer of similar quality
Specifications
Detectable AnalytesPlatelets
For Use With (Equipment)AggRAM
Sample TypePlasma
IncludesADP reagent 2 x 1 mL, Collagen Reagent, 2 x 1 mL, Epinephrine Reagent, 2 x 1 mL
Unit SizeEA
Contents & Storage
2°C to 8°C