Check Order Status
Epinephrine
Actual product may vary
Epinephrine
Epinephrine is for use in routine platelet aggregation studies for evaluation of qualitative platelet function disorders
Have Questions?
Catalog Number
Quantity
TYL5367
2 x 1 mL Vials
Catalog number
TYL5367
Price (MYR)
956.00
EA
Add to cart
Quantity:
2 x 1 mL Vials
Price (MYR)
956.00
EA
Add to cart
Epinephrine is for use in routine platelet aggregation studies for evaluation of qualitative platelet function disorders
Reagents are optimized for use with AggRAM, but may be used on any turbidometric aggregometer of similar quality
Epinephrine is a lyophilized preparation of L-epinephrine bitartrate
Epinephrine is for use in routine platelet aggregation studies for evaluation of qualitative platelet function disorders
After reconstitution, epinephrine concentration is 3 mM
Specifications
Detectable Analytes
Platelets
For Use With (Equipment)
AggRAM
Sample Type
Plasma
Quantity
2 x 1 mL Vials
Form
Lyophilized
Unit Size
EA
Contents & Storage
2°C to 8°C