Check Order Status
Tempered Cast Iron Bossheads
Actual product may vary
Tempered Cast Iron Bossheads
Manufactured from malleable cast iron (DIN 12895). Bochem™ Tempered Cast Iron Bossheads feature thumb and safety screws.
Have Questions?
Catalog Number
Diameter (Metric) Outer
Product Type
TYL5360
16.5 mm
Bosshead
Catalog number
TYL5360
Price (MYR)
956.00
EA
Add to cart
Diameter (Metric) Outer:
16.5 mm
Product Type:
Bosshead
Price (MYR)
956.00
EA
Add to cart
Malleable cast iron (DIN 12895)
Type 1: Thumb screw M8
Type 4: Safety screw type
heart
M8
Specifications
Material
Tempered Cast Iron
Diameter (Metric) Outer
16.5 mm
Product Type
Bosshead
Thread Size
M8
Description
Screw Type: 1 Thumb screw M8
Quantity
1 Ea.