Check Order Status
Double-Sleeve Malleable Cast Iron Bosshead
Actual product may vary
Double-Sleeve Malleable Cast Iron Bosshead
Manufactured from malleable cast iron. BOCHEM™ Double-Sleeve Malleable Cast Iron Bosshead is DIN 12895 certified.
Have Questions?
Catalog Number
Diameter (Metric)
Material
Type
TYL5359
20 mm
Malleable Cast Iron
Double Sleeve
Catalog number
TYL5359
Price (MYR)
956.00
EA
Add to cart
Diameter (Metric):
20 mm
Material:
Malleable Cast Iron
Type:
Double Sleeve
Price (MYR)
956.00
EA
Add to cart
Thumb screw M8
Malleable cast iron (DIN 12895)
Chromium plated
Specifications
Quantity
1 Ea.
Type
Double Sleeve
Material
Malleable Cast Iron
Diameter (Metric)
20 mm
Diameter (Metric) Inside
20 mm