Aluminum Bosshead
Aluminum Bosshead
Actual product may vary
Aluminum Bosshead
Aluminum Bosshead
Aluminum Bosshead
Bochem™ Aluminum Bosshead features an M8 heart type safety screw with a 90° and 180° angle.
Have Questions?
Catalog NumberSize
TYL53568 mm thread
Catalog number TYL5356
Price (MYR)
956.00
EA
Add to cart
Size:
8 mm thread
Price (MYR)
956.00
EA
Add to cart
  • Aluminum
  • M8 heart type screw
Specifications
MaterialAluminum
DescriptionM8 heard safety screw
Diameter (Metric)16.5 mm
Size8 mm thread
Unit SizeEA