Stainless Steel Bosshead
Stainless Steel Bosshead
Actual product may vary
Stainless Steel Bosshead
Bochem™ bossheads are manufactured from 18/10 stainless steel, with 180° and 90° angle, and M8 'heart' safety screw.
Have Questions?
Catalog NumberColor
TYL5354Silver
Catalog number TYL5354
Price (MYR)
956.00
EA
Add to cart
Color:
Silver
Price (MYR)
956.00
EA
Add to cart
  • 18/10 stainless steel
  • Safety screw type 'heart' M8
  • 180+90° angle
Specifications
TypeBosshead Clamp
MaterialStainless Steel
IncludesSafety Screw Type 'Heart' M8
For Use With (Equipment)Rods and Stand Bases
For Use With (Application)Holding and Supporting Laboratory Apparatus
Diameter (Metric)16.5 mm
ColorSilver
Unit SizeEA