U.S. Standard Stainless Steel Sieves, 8 in. dia. x 2 in.D
U.S. Standard Stainless Steel Sieves, 8 in. dia. x 2 in.D
Actual product may vary
U.S. Standard Stainless Steel Sieves, 8 in. dia. x 2 in.D
Stainless Steel. Aperture sizes from 5.00 in. opening to 500 mesh.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPore Size
TYL521375 μm
FIS04-881-10G2 mm
TYL51942 mm
TYL51971.18 mm
FIS04-881-10L1 mm
TYL5206250 μm
FIS04-881-10Z106 μm
FIS04-881-10BB75 μm
Catalog number TYL5213
Price (MYR)
1,298.00
EA
Add to cart
Pore Size:
75 μm
Price (MYR)
1,298.00
EA
Add to cart
Stainless Steel. Aperture sizes from 5.00 in. opening to 500 mesh.
All sieves are serialized and come with a Certificate of Compliance.
If Test Certificate is required (Fisher Cat. No. 04-909-16), it must be orderd along with the sieve.
The list below does not represent the complete ASTM E 11 specifications; other sizes are available. Contact your Fisher representative for more information.
Specifications
Depth (English)2 in.
Depth (Metric)5.08 cm
Pore Size75 μm
TypeFull Height Test Sieve
Certifications/ComplianceASTM E11-09, ISO 565 3310-1
Diameter (English) Outer8 in.
MaterialStainless Steel
Height (English)2.62 in.
IncludesTest sieve certificate
Height (Metric)6.65 cm
Tyler Standard200 Mesh
U.S. StandardNo. 200
For Use With (Application)Ideal for particle measurement in agricultural, bio industry, chemical, cosmetics, general industrial, fertilizer, foods, minerals, pharmaceuticals, Plastics and Metal products industries
Diameter (Metric) Outer203 mm
Frame MaterialStainless Steel
Screen Size200 Mesh
Mesh MaterialStainless Steel
Sieve Number200
Unit SizeEA