Glass pH Electrode
Glass pH Electrode
Actual product may vary
Glass pH Electrode
Have Questions?
Catalog NumberMaterial
TYL5209Glass
Catalog number TYL5209
Price (MYR)
984.00
EA
Add to cart
Material:
Glass
Price (MYR)
984.00
EA
Add to cart
Specifications
MaterialGlass
pH Range0 to 14
Temperature (Metric) Operating0°C to 80°C
TypepH Electrode
For Use With (Application)In biology and clinical.
Temperature (Metric) Range0°C to 80°C
Unit SizeEA