52 02 pH Electrode
52 02 pH Electrode
Actual product may vary
52 02 pH Electrode
The 52 02 is a pH electrode with screw cap S7, for general use in the laboratory. It features a large, semi-spheric membrane for very robust and fast responding.
Have Questions?
Catalog NumberConnector Types
TYL5202S7
Catalog number TYL5202
Price (MYR)
956.00
EA
Add to cart
Connector Types:
S7
Price (MYR)
956.00
EA
Add to cart
  • Average life expectancy is approx. one year, depending on the working conditions
  • Measuring pH range from 0 to 12
  • Operating temperature from 0° to 80°C
  • Reference element of Ag wire coated with AgCl
  • Ceramic diaphragm and CRISOLYT A electrolyte
  • Min. immersion depth of 20mm
  • Applications include aqueous media in general
  • Limitations include low-conductivity, dirty or viscous solutions, products with colloids or solids in suspension, samples containing sulphides or reducing substances, and highly alkaline samples
Specifications
Connector TypesS7
MaterialGlass
pH Range0 to 12
For Use With (Application)Aqueous media in general
MembraneHigh
Model52 02
Temperature (Metric) Range0°C to 80°C
Unit SizeEA