Ristocetin for Aggregation Assays
Ristocetin for Aggregation Assays
Actual product may vary
Ristocetin for Aggregation Assays
Ristocetin is for use in platelet aggregation studies for the confirmation of von Willebrand Disease (vWD) and Bernard-Soulier Syndrome.
Have Questions?
Catalog NumberQuantity
TYL519910 x 0.5 mL Vials
Catalog number TYL5199
Price (MYR)
956.00
EA
Add to cart
Quantity:
10 x 0.5 mL Vials
Price (MYR)
956.00
EA
Add to cart
Ristocetin is for use in platelet aggregation studies for the confirmation of von Willebrand Disease (vWD) and Bernard-Soulier Syndrome.
  • Reagents are optimized for use with AggRAM, but may be used on any turbidometric aggregometer of similar quality
  • This Ristocetin is for use in platelet aggregation studies for confirmation of von Willebrand Disease (vWD) and Bernard-Soulier Syndrome
  • Normal platelet-rich plasma exhibits a positive aggregation response to the addition of ristocetin using a final concentration of 12 to 15 mg/mL
  • The majority of vWD patients exhibit a negative response to ristocetin as do Bernard-Soulier patients.
Specifications
Concentration12 mg/mL
Detectable AnalytesRistocetin
FormatVial
For Use With (Equipment)AggRAM
TypeLyophilized
Quantity10 x 0.5 mL Vials
Unit SizeEA
Contents & Storage
2°C to 8°C