Factor VIII Deficient Substrate Plasma
Factor VIII Deficient Substrate Plasma
Actual product may vary
Factor VIII Deficient Substrate Plasma
Factor VIII Deficient Substrate Plasma is intended for the quantitative determination of Factor VIII (antihemophilic factor) in patients suspected of having congenital or acquired deficiency of this coagulation protein
Have Questions?
Catalog NumberQuantity
TYL51931 mL
Catalog number TYL5193
Price (MYR)
956.00
EA
Add to cart
Quantity:
1 mL
Price (MYR)
956.00
EA
Add to cart
Factor VIII Deficient Substrate Plasma is intended for the quantitative determination of Factor VIII (antihemophilic factor) in patients suspected of having congenital or acquired deficiency of this coagulation protein
  • Factor VIII Deficient Substrate is used for quantitative determination of Factor VIII in patients suspected of having congenital or acquired deficiency of this coagulation protein
  • Available as a lyophilized product in 10 x 1mL vials.
Specifications
DescriptionStore at 2 to 6°C
Detectable AnalytesAntihemophilic factor
Packaging Type10 Vials
Quantity1 mL
Unit SizeEA