Metzenbaum Scissors
Metzenbaum Scissors
Actual product may vary
Metzenbaum Scissors
Precisely cuts tough or delicate tissue
Have Questions?
Catalog NumberLength (Metric)Blade StyleProduct Type
TYL518117.78 cmStraightMetzenbaum Scissors
Catalog number TYL5181
Price (MYR)
956.00
EA
Add to cart
Length (Metric):
17.78 cm
Blade Style:
Straight
Product Type:
Metzenbaum Scissors
Price (MYR)
956.00
EA
Add to cart
  • High-quality surgical-grade stainless steel construction provides optimal performance
  • Fully tempered precision screw locks maintain perfect alignment
  • Ergonomic design combines weight, balance and feel for optimal comfort and performance
  • Well rounded and polished gold ring handles provide exceptional comfort and reduced hand fatigue
  • Straight or curved blades
  • Blunt points
  • Tungsten carbide inserts seamlessly bonded to upper and lower blades
  • SuperCut scissors have a specially designed razor sharp edge that cuts effortlessly through tissue; lower blade has micro fine serrations to prevent tissue slippage
Specifications
Product TypeMetzenbaum Scissors
Blade StyleStraight
GradePremium
Length (English)7 in.
Length (Metric)17.78 cm
MaterialStainless Steel
For Use With (Application)Surgical, metzenbaum
Unit SizeEA