Sentino™ Microbiology Pump Replacement Adapter
Sentino™ Microbiology Pump Replacement Adapter
Actual product may vary
Sentino™ Microbiology Pump Replacement Adapter
Pump replacement adapter for use with MicroFunnel filter funnels.
Have Questions?
Catalog number TYL4872
Price (MYR)
849.00
EA
Add to cart
Price (MYR)
849.00
EA
Add to cart
  • For vacuum filtration of liquids
  • For the analysis of aqueous samples for microbiological and particulate contamination using the Membrane Filter (MF) Technique
  • For research use only
Specifications
For Use With (Equipment)MicroFunnel™ Filter Funnel
MaterialStainless Steel (316L)