Check Order Status
Sulphuric acid in aqueous solution, DILUT-IT™, J.T. Baker™
Element Search
Structure Search
Sulphuric acid in aqueous solution, DILUT-IT™, J.T. Baker™
Laboratory Reagent
Have Questions?
Catalog number
TYL4869
Price (MYR)
849.00
Price (MYR)
849.00
EA
Add to cart
Specifications
CAS
7664-93-9
Concentration
0.005 mol
Density
1.01 g/cm³ (20°C)
Recommended Storage
Ambient
Quantity
50 mL
View more