Acrodisc™ Syringe Filters with PTFE Membrane and Male Slip Luer Outlet
Acrodisc™ Syringe Filters with PTFE Membrane and Male Slip Luer Outlet
Actual product may vary
Acrodisc™ Syringe Filters with PTFE Membrane and Male Slip Luer Outlet
Universal membrane syringe filters for all of your analytical filtration requirements.
Have Questions?
Catalog NumberPore SizeQuantity
TYL45430.45 μm1000/Pk.
Catalog number TYL4543
Price (MYR)
849.00
EA
Add to cart
Pore Size:
0.45 μm
Quantity:
1000/Pk.
Price (MYR)
849.00
EA
Add to cart
  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Material (Membrane)PTFE
Pore Size0.45 μm
SterilityNon-sterile
Filtration Area1 cm2
Hold up Volume (After Air Purge)<30 μL
Housing MaterialPolypropylene
Outlet StyleMale Slip Luer Outlet
Quantity1000/Pk.
Temperature (Metric) Max100°C
Diameter (Metric)13 mm
Unit SizeEA