Acrodisc™ Syringe Filters with Nylon Membrane and Male Slip Luer Outlet
Acrodisc™ Syringe Filters with Nylon Membrane and Male Slip Luer Outlet
Actual product may vary
Acrodisc™ Syringe Filters with Nylon Membrane and Male Slip Luer Outlet
Versatile filter for both aqueous and solvent-based sample filtration.
Have Questions?
Catalog NumberPore SizeQuantity
TYL45410.45 μm1000/Pk.
Catalog number TYL4541
Price (MYR)
849.00
EA
Add to cart
Pore Size:
0.45 μm
Quantity:
1000/Pk.
Price (MYR)
849.00
EA
Add to cart
  • Excellent chemical compatibility with esters, bases, and alcohols
  • Saves time because no prewetting is required
Specifications
Material (Membrane)Nylon
Pore Size0.45 μm
SterilityNon-sterile
Filtration Area1 cm2
Hold up Volume (After Air Purge)<30 μL
Housing MaterialPolypropylene
Outlet StyleMale Slip Luer Outlet
Quantity1000/Pk.
Temperature (Metric) Max55°C
Diameter (Metric)13 mm
Unit SizeEA