Brass Test Sieves, 8 in. dia. x 2 in. D
Brass Test Sieves, 8 in. dia. x 2 in. D
Actual product may vary
Brass Test Sieves, 8 in. dia. x 2 in. D
Brass. Aperture sizes from 5.00 in. opening to 500 mesh.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSieve Number
TYL453940
TYL453625
Catalog number TYL4539
Price (MYR)
849.00
EA
Add to cart
Sieve Number:
40
Price (MYR)
849.00
EA
Add to cart
Brass. Aperture sizes from 5.00 in. opening to 500 mesh.
  • All sieves are serialized.
  • The list below does not represent the complete ASTM E 11 specifications; other sizes are available.
Note:
A basic certificate of compliance comes with every test sieve. Additional certification is offered for customers who have more stringent QC programs; contact Thermo Fisher Customer Service.
Specifications
Depth (English)2 in.
Depth (Metric)5.08 cm
Pore Size425 μm
TypeFull Height Test Sieve
Diameter (English) Outer8 in.
MaterialBrass
Height (English)2.62 in.
IncludesTest sieve certificate
Height (Metric)6.65 cm
Tyler Standard35 Mesh
U.S. StandardNo. 40
For Use With (Application)Ideal for particle measurement in agricultural, bio industry, chemical, cosmetics, general industrial, fertilizer, foods, minerals, pharmaceuticals, Plastics and Metal products industries
Diameter (Metric) Outer203 mm
Frame MaterialBrass
Screen Size40 Mesh
Mesh MaterialBrass
Sieve Number40
Unit SizeEA