Elplasia™ 12K Flask
Elplasia™ 12K Flask
Actual product may vary
Elplasia™ 12K Flask
The Elplasia™ 12K Flask enables researchers to generate a high density of animal-free spheroids in a scaffold-free model.
Have Questions?
Catalog number TYL4537
Price (MYR)
849.00
EA
Add to cart
Price (MYR)
849.00
EA
Add to cart
  • Contains 152 microcavities per cm2 in a vessel footprint similar to that of a T-75 flask
  • Corning™ Ultra-Low Attachment (ULA) surface works with gravity and a rounded microcavity geometry to form approximately 12,000 spheroids of similar shape and size
  • ULA surface (a proprietary, animal-free, covalently bonded hydrogel), promotes the formation and easy harvesting of spheroids
  • Polystryene film is gas permeable with low autofluorescence
  • Round well-bottom geometry
Specifications
Closure ColorOrange
For Use With (Application)Spheroid Formation
Bottom ShapeRectangular
Culture Area80 cm2
Volume (Metric) Working25 to 50 mL
Unit SizeEA