Combination Wrench, 13mm
Combination Wrench, 13mm
Actual product may vary
Combination Wrench, 13mm
Ampco Safety Tools Combination Wrench, 13mm is non-sparking, non-magnetic and corrosion resistant
Have Questions?
Catalog NumberLength (Metric)
TYL1310204.78 mm
Catalog number TYL1310
Price (MYR)
769.00
EA
Add to cart
Length (Metric):
204.78 mm
Price (MYR)
769.00
EA
Add to cart
Made in USA
Specifications
For Use With (Equipment)Electronics, batteries, MRI, and precision magnetic equipment
Length (Metric)204.78 mm
MaterialAluminum Bronze
ColorBronze
For Use With (Application)Non-Sparking, Non-Magnetic, Corrosion Resistant Applications; Flammable or combustible vapors, liquids, dusts or residues; Corrosive environments like laboratories, acid, salt water and chemical processing
No. per Pack1 Ea.
Weight (Metric)90 g
Width (Metric)34.92 mm
Unit SizeEA