Check Order Status
Stainless Steel Wire Basket for Vertical Autoclaves
Actual product may vary
Stainless Steel Wire Basket for Vertical Autoclaves
Stainless steel mesh wire basket for bottles and general use.
Have Questions?
Catalog Number
For Use With (Equipment)
Dimensions (L x W)
TTLBSK507-0003
5075 Vertical Autoclave
465 x 235 mm
Catalog number
TTLBSK507-0003
Price (MYR)
-
For Use With (Equipment):
5075 Vertical Autoclave
Dimensions (L x W):
465 x 235 mm
Stainless Steel wire basket
Specifications
Description
Maximum number of baskets: 3
Capacity (English)
8.8 gal.
Capacity (Metric)
40 L
Depth (Metric) Chamber
23.5 cm
Dimensions (L x W)
465 x 235 mm
Depth (English) Chamber
9.4 in.
For Use With (Equipment)
5075 Vertical Autoclave
Length (English)
18.3 in.
Length (Metric)
46.5 cm
Product Type
Autoclave Basket
Sterility
Non-sterile