Isothermal/Disinfection (for Agar Preparation)
Isothermal/Disinfection (for Agar Preparation)
Actual product may vary
Isothermal/Disinfection (for Agar Preparation)
Isothermal/Disinfection (for Agar preparation). The isothermal cycle has a temperature range from 60°C to 94°C and allows for gentle heating and cooling of agar.
Have Questions?
Catalog number TTLADD222-0447
Price (MYR)
-
  • Disinfection (agar prep)
  • Isothermal/Disinfection (for Agar preparation).
  • The isothermal cycle has a temperature range from 60°C to 94°C and allows for gentle heating and cooling of agar.
Specifications
For Use With (Application)Agar Preparation
Product TypeSoftware
Unit SizeEA