Front Loading Bench Top Autoclave
Front Loading Bench Top Autoclave
Actual product may vary
Front Loading Bench Top Autoclave
Front Loading Bench Top Autoclave
Front Loading Bench Top Autoclave
Each model has a number of optional added-value features
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Includes
TTL3850EL-D65 L2 x PT100 Temp Probe
TTL2840EL-D28 L2 x PT100 Temp Probe
Catalog number TTL3850EL-D
Price (MYR)
91,608.00
EA
Add to cart
Capacity (Metric):
65 L
Includes:
2 x PT100 Temp Probe
Price (MYR)
91,608.00
EA
Add to cart
For over 90 years, Tuttnauer has been an industry leader satisfying customer expectations with top quality, high-performance products and a dedicated service support team. The laboratory autoclave line is designed for laboratory applications used in research institutes, universities, medical, pharmaceutical, biotechnology, food and chemical industries. Each model has a number of optional added-value features which can be configured for fast cooling, efficient drying, biohazard, waste sterilization and more. Benchtop lab autoclaves are designed to save space on your laboratory workbench.
  • Fast Liquid Cooling
    After sterilization is completed, compressed air is passed through a microbiological filter into the autoclave chamber in order to prevent a drop in pressure which also prevents load deformation, cracks or spills. Cold water is then circulated through cooling pipes that rapidly reduces the chamber temperature and that of the liquid load to a safe temperature. Tuttnauer’s fast liquid cooling technology reduces cycle time by as much as 75% and minimizes load exposure to high temperatures.
  • Super Fast Liquid Cooling
    In addition to fast cooling, an optional fan can be applied to further circulate the compressed air in the chamber. This speeds up the heat exchange during the cooling stage in order to safely achieve super fast cooling of the liquid load under pressure. Tuttnauer’s accelerated fast liquid cooling technology reduces cycle time by as much as 90% and minimizes load exposure to high temperatures.
  • F0 – Protect Your Liquid Media, Save Time, Save Energy
    An additional challenge with liquid sterilization is the need to prevent extended exposure of liquid media to high temperatures which may harm the quality of the liquid media. The advanced F0 optional feature assists in minimizing the time liquids are exposed to high temperatures during sterilization thereby protecting liquid media, saving your laboratory time and reducing energy consumption.
  • Fast and Efficient Heat-up
    Vertical and Benchtop laboratory autoclaves with built-in steam generators provide fast & efficient heat-up and complete drying.
  • Efficient Air Removal
    An optional vacuum pump can be used for fractioned pre vacuum air removal eliminating air pockets from all load types and maximizing efficient steam penetration throughout the entire load.
  • Active Drying with Post Vacuum
    For benchtop autoclaves, an optional vacuum pump can be used for post vacuum drying, at the end of the sterilization cycle, ensuring improved drying of porous loads and hollow instruments such as pipette tips. The benchtop autoclave is equipped with a heating plate attached under the chamber that heats the chamber during the drying phase. The low pressure in the autoclave chamber, caused by the vacuum, reduces the boiling temperature forcing moisture to evaporate rapidly. The vapour is then removed from the chamber by vacuum resulting in a dry load.
  • R.PC.R Software
    • Automatic Recording of Cycle Information to Your PC
    • Reporting You Can Rely On
    • Remote PC Reporting (optional PC software)
    • Automatic recording of cycle information to any PC on your network
    • Convenient access to graphs and tables that are easy to understand
    • Easily generate PDF reports
    • No need to file printouts, saving you time
    • Be in Control with Real-Time Remote Monitoring
    • See the real-time autoclave display on your PC
    • Monitor all activity for up to 8 autoclaves
  • Advanced Control System for Your Laboratory
    Take Advantage of Tuttnauer’s state-of-the-art Control System with Multi-Color Display. Features:
    • Fo software control
    • PID (Proportional Integral Differential) pressure control
    • Stores the last 200 cycles in built-in memory
    • Two independent flexible PT100 temperature sensors to prevent over-boiling of liquids and explosions of bottles
    • Up to 6 temperature sensors and 4 pressure sensors can be connected
    • 30 Identification Codes and Passwords for access level control
    • The controller and software comply with the 21 CFR part 11 standard
  • Innovative Multi-Color Display
    • Multi-color display for easier reading
    • Color is used to indicate the stage of the cycle
    • Quick access to important information
    • 26 Languages
    • Built-in view of historical cycle data
Specifications
Controller TypeMicroprocessor
Amperage15 A
Capacity (English)68.7 qt.
Capacity (Metric)65 L
Depth (Metric) Chamber500 mm
Dimensions (L x W)Chamber: 380 x 500 mm
Diameter (English) Chamber15.0 in.
Depth (English) Chamber19.7 in.
Diameter (Metric) Chamber380 mm
For Use With (Application)Steam sterilization
For Use With (Equipment)Based on Configuration, a SS Basket, SS Container or Loading Cart and Trolley May be Recommended
Length (English)30.1 in.
Height (English) Exterior21.3 in.
Height (Metric) Exterior540 mm
Length (Metric)765 mm
ModelFront-loading
Product TypeBenchtop Autoclave
Electrical Requirements40 V 50 Hz 15 A Single Phase
FeaturesInternal Reservoir
Includes2 x PT100 Temp Probe
Width (Metric) Exterior720 mm
Dimensions (L x W x H)765 x 720 x 540 mm
Unit SizeEA