TSX Series High-Performance Undercounter Lab Refrigerators
TSX Series High-Performance Undercounter Lab Refrigerators
Actual product may vary
TSX Series High-Performance Undercounter Lab Refrigerators
TSX Series High-Performance Undercounter Lab Refrigerators
TSX Series High-Performance Undercounter Lab Refrigerators
TSX Series High-Performance Undercounter Lab Refrigerators
For a better environment inside and out - Introducing our new undercounter lab refrigerator.
Have Questions?
Catalog NumberDoor StylePlug Type
TSX505SASingle SolidNEMA 5-15 P
Catalog number TSX505SA
Price (MYR)
50,314.00
EA
Add to cart
Door Style:
Single Solid
Plug Type:
NEMA 5-15 P
Price (MYR)
50,314.00
EA
Add to cart
Thermo Scientific™ TSX Series high-performance undercounter lab refrigerators are designed with features that support sample protection and sustainability objectives for the storage of media, reagents, pharmaceuticals, vaccines, chemotherapy, and other medical and pharmacy-grade storage requiring 2° to 8°C. The Series' V-drive is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.
Unique to the TSX505, our dual refrigeration solution operates through the action of two synchronized solid-state heat pumps. The two solid-state heat pumps work to cool a combination of cold wall and forced air to help ensure that conditions are optimal for the most sensitive applications
Certified to NSF/ANSI 456 Vaccine Storage standard, the TSX series can help you meet regulations and guidelines from the CDC, World Health Organization, and other Health Ministries on vaccine storage and handling.
The TSX Series delivers the performance and quality you need, in addition to the low sound, low energy, and exceptional user experience you deserve.
The Drive for Sample Integrity and Security
  • Designed to safely store media, reagents, pharmaceuticals, vaccines, chemotherapy and other medical-grade storage requiring a temperature range of 2°C to 8°C
  • Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection
  • Redundant cooling offers additional peace of mind for your most precious samples
  • Automated electronic passcode locking standard
  • Datalogging with USB download functionality
Enabling Compliance
  • Tested and Certified to NSF/ANSI 456 Vaccine Storage standard and supports the US CDC, WHO, and other global vaccine storage guidelines and recommendations
  • Tested to GMP Clean Room Class A / ISO 7 (ISO EN 14644-1) with easy to clean surfaces support USP <797> and <800> hazardous drug compounding and other clean room environments
  • Tested and certified to conform with the design and safety standards of UL 61010 and CE Directive 2011/65/EU and 2015/863
  • ADA compliant
Designed for an Ergonomic and Comfortable Experience
  • Ergonomically designed door handle
  • Touchscreen controller with password protection is angled to help prevent neck strain
  • Whisper quiet with a sound level of 35 dBA, enabling end users to work alongside the refrigerator without distraction
  • Bright, easy-to-adjust interior LED lighting
  • Self-closing door with stop to assist with inventory loads
  • Universal voltage (100-240v, 50/60hz) allows placement throughout the facility and globally just by connecting the region-specific power cord.
Verified Sustainability
  • US EPA ENERGY STAR Certified with up to 70% less energy usage than conventional refrigerant models
  • Natural, CO2 and Water refrigerants and green foam make the TSX Series compliant with EU F-GAS regulations, U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP), and other Global Environmental Regulations.
Ready for Connectivity and Monitoring
  • Compatible with wireless monitoring systems and remote alarm contacts including our Thermo Scientific™ Smart-Vue™ Pro wireless monitoring systems
Make It Your Own
  • Touchscreen controller with password protection
  • Choice of reversible glass or solid door
  • Extra and Alternate wire baskets and shelves
  • Specialty, regional, and country specific plugs
Services and Support
  • Included Certificate of Calibration to an NIST (National Institute of Standards and Technology, formerly National Bureau of Standards) traceable instrument includes a model number and serial number certificate
  • Optional Validation Testing Programs include in house and field-based temperature mapping and Installation Qualification (IQ) and Operation Qualification (OQ) programs
Specifications
DescriptionTSX Series High-Performance Undercounter Refrigerator, Solid Door
TypeHigh-Performance Refrigerator
Capacity (English)5.5 cu. ft.
Capacity (Metric)156 L
Temperature Range3°C to 7°C
RefrigerantR744
Door StyleSingle Solid
No. of Doors1
Shelves3
Certifications/ComplianceCE, REACH, RoHS, UL
StyleUndercounter
DisplayFull color TFT-LCD; 4.3”; 480 x 272
Product LineTSX Series
DefrostAutomatic
Monitoring OptionsAlarm
Smart View CompatibilitySmart-Vue Compatible
Dimensions (D x W x H) Interior20.5 x 19.5 x 23 in.(52 x 49.5 x 58.4 cm)
Dimensions (L x W x H) Exterior26 x 23.6 x 31.8 in. (66 x 60 x 81 cm)
Depth (English) Exterior26 in.
Depth (English) Interior20.5 in.
Depth (Metric) Exterior66 cm
Depth (Metric) Interior52 cm
Height (English) Exterior31.8 in.
Height (English) Interior23 in.
Height (Metric) Exterior79.5 cm
Height (Metric) Interior58.4 cm
Width (English) Exterior23.6 in.
Width (English) Interior19.5 in.
Width (Metric) Exterior60 cm
Width (Metric) Interior49.5 cm
Amperage10 A Max / 3 A Steady State
Plug TypeNEMA 5-15 P
Voltage100/240 V
Electrical Requirements100 - 240 V 50-60 Hz
Frequency50/60 Hz
Green FeaturesEnergy efficient
Shipping Weight (English)155 lb.
Shipping Weight (Metric)71 kg
Weight (English)119 lb.
Weight (Metric)53.98 kg
Holds45 Boxes per Level (9 x 5 Pattern), 225 Boxes per Shelf (5 Levels per Shelf, 2 Shelves per Unit), +45 Boxes (1 Additional Level Above Top Shelf)
Unit SizeEA