Search Thermo Fisher Scientific
| Catalog Number | Quantity |
|---|---|
| TSSDC-20 | 20/Pk. |
Steriking™ daily control B&D Test Pack pk20
The STERIKING™ Daily Control test packs are single-service Bowie & Dick type tests for pre-vacuum sterilizers for steam penetration, steam quality and air leak detection. The STERIKING™ Daily Control Test packs are designed to monitor the attainment of the critical variables in the Bowie & Dick test program.