TSE Series Solid Door Laboratory Refrigerators
TSE Series Solid Door Laboratory Refrigerators
Actual product may vary
TSE Series Solid Door Laboratory Refrigerators
TSE Series Solid Door Laboratory Refrigerators
TSE Series Solid Door Laboratory Refrigerators
TSE Series Solid Door Laboratory Refrigerators
TSE Series Solid Door Laboratory Refrigerators
Digital control and temperature alarms deliver dependable storage for everyday supplies. Thermo Scientific Laboratory Refrigerators solid door models offer consistent performance for routine lab use in convenient undercounter and upright single door sizes that fit a variety of laboratory spaces.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
TSE2205SV564 L
TSE0405SV77 L
TSE1205SV266 L
Catalog number TSE2205SV
HSN 84184090
Price (MYR)
29,326.00
Each
Add to cart
Capacity (Metric):
564 L
Price (MYR)
29,326.00
Each
Add to cart
Thermo Scientific TSE Series Solid Door Laboratory Refrigerators provide dependable temperature performance and straightforward operation for routine lab use. Digital control and essential features support daily storage needs, and solid doors help protect contents from ambient light. All models use SNAP and F-Gas compliant hydrocarbon refrigerants and include a two-year warranty, offering practical value for everyday laboratory workflows.
  • Designed for everyday research and academic lab applications with good performance and easy-to-use digital controls
  • Quiet operation, less than 53 dB
  • SNAP compliant green refrigerants
  • Auto defrost
  • Four 2 in. casters, front casters are lockable
  • Hi/Lo Temperature, and door ajar failure alarms
  • Rear access port
  • Keyed door locks, compatible with popular E-lock adapters
Specifications
Capacity (English)20 cu.ft.
Capacity (Metric)564 L
Temperature Range1°C to 10°C
Temperature (Setpoint)5°C
RefrigerantR290
Door StyleSolid
No. of Doors1
Shelves4
Cabinet MaterialPainted Steel
Exterior FinishWhite-coated Steel
InsulationHFC-free Polyurethane Foam
Certifications/ComplianceCE
StyleUpright
DisplayDigital
CastersLocking
Control TypeDigital
Ports1 Rear-access Port
Warranty2 Years Parts & Labor
DefrostAuto
LightingLED
Monitoring OptionsAudible and Visual High and Low Temperature Alarms
Access SecurityKey Lock
CompressorHermetic
Dimensions (DxWxH) Exterior34.7 x 28.1 x 79.7 in. (88.1 x 71.3 x 202.6 cm)
Dimensions (D x W x H) Interior27.6 x 23.5 x 59.3 in. (70.0 x 59.6 x 150.5 cm)
Plug TypeEU
Voltage230 V
Frequency50 Hz
Energy Usage1.48 kw-hr/day
Shipping Weight (English)317 lb.
Shipping Weight (Metric)144 kg
Unit SizeEach